missing translation for 'onlineSavingsMsg'
Få mere at vide

Quetiapine Dimer Impurity, TRC

Product Code. 30420168
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30420168 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420168 Supplier Toronto Research Chemicals Supplier No. TRCQ5099605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Quetiapine Dimer Impurity
CAS 945668-94-0
Molekylær formel C30 H24 N4 S2
Mængde 5 mg
Synonym Dibenzo[b,f][1,4]thiazepine','11,11'-(1,4-piperazinediyl)bis-, 11,11'-(1,4-Piperazinediyl)bis-dibenzo[b,f][1,4]thiazepine, 11,11'-(1,4-Piperazinediyl)bis[dibenzo[b,f][1,4]thiazepine], Quetiapine Fumarate Imp. D (EP), Quetiapine dimer impurity, Bis(dibenzothiazepinyl) piperazine, 1,4-Bis(dibenzo[b,f][1,4]thiazepin-11-yl)piperazine
InChI formel InChI=1S/C30H24N4S2/c1-5-13-25-21(9-1)29(31-23-11-3-7-15-27(23)35-25)33-17-19-34(20-18-33)30-22-10-2-6-14-26(22)36-28-16-8-4-12-24(28)32-30/h1-16H,17-20H2
SMIL C1CN(CCN1C2=Nc3ccccc3Sc4ccccc24)C5=Nc6ccccc6Sc7ccccc57
IUPAC navn 6-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)benzo[b][1,4]benzothiazepine
Molekylvægt (g/mol) 504.67
Formel vægt 504.1442
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.