Få mere at vide
Quetiapine-d8 Hemifumarate (piperazine-d8), TRC
Artikelnummer. 30423559
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30423559 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423559 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ51000610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Quetiapine-d8 Hemifumarate (piperazine-d8)
CAS 1435938-24-1
Molekylær formel 2 C21 H17 D8 N3 O2 S . C4 H4 O4
Mængde 10 mg
InChI formel InChI=1S/2C21H25N3O2S.C4H4O4/c2*25-14-16-26-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)27-20-8-4-2-6-18(20)22-21;5-3(6)1-2-4(7)8/h2*1-8,25H,9-16H2;1-2H,(H,5,6)(H,7,8)/b;;2-1+/i2*9D2,10D2,11D2,12D2;
SMIL [2H]C1([2H])N(CCOCCO)C([2H])([2H])C([2H])([2H])N(C2=Nc3ccccc3Sc4ccccc24)C1([2H])[2H].[2H]C5([2H])N(CCOCCO)C([2H])([2H])C([2H])([2H])N(C6=Nc7ccccc7Sc8ccccc68)C5([2H])[2H].OC(=O)\C=C\C(=O)O
IUPAC navn 2-[2-(4-benzo[b][1,4]benzothiazepin-6-yl-2,2,3,3,5,5,6,6-octadeuterio-piperazin-1-yl)ethoxy]ethanol;fumaric acid
Molekylvægt (g/mol) 899.18495
Formel vægt 898.4449