Få mere at vide
Quercetin 3-O-Beta-D-Glucuronide, TRC
Artikelnummer. 30003784
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003784 5 mg
30005517 10 mg
30006224 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003784 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ5095105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22688-79-5
Molekylær formel C21 H18 O13
Molekylvægt (g/mol) 478.36
Synonym 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-oxo-4H-1-benzopyran-3-yl β-D-Glucopyranosiduronic Acid, Miquelianin, Mikwelianin, Quercetin 3-O-Glucuronide, Quercetin 3-O-β-D-Glucuronide, Quercetin 3-O-β-D-Glucuronopyranoside, Quercetin 3-Glucuronide, Quercetin 3-β-Glucuronide, Quercetin Glucuronide, Quercetin β-Glucuronide, Quercituron, Querciturone;
IUPAC navn (2S,3S,4S,5R)-6-[2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4-oxochromen-3-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL O[C@@H]1[C@@H](O)C(OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)O[C@@H]([C@H]1O)C(=O)O
Tekniske data
Kemisk navn eller materiale Quercetin-3-O-glucuronide
Mængde 5 mg
InChI formel InChI=1S/C21H18O13/c22-7-4-10(25)12-11(5-7)32-17(6-1-2-8(23)9(24)3-6)18(13(12)26)33-21-16(29)14(27)15(28)19(34-21)20(30)31/h1-5,14-16,19,21-25,27-29H,(H,30,31)/t14-,15-,16+,19-,21?/m0/s1
Formel vægt 478.0747