Learn More
Miele™ Quarter Segment Test Tube Insert
Brand:  Miele™ 6907660
Additional Details : Weight : 50.00000kg
 View more versions of this product
Product Code. 13102270
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Quarter Segment Test Tube Insert
320 mm
200 mm
Test tubes up to 12 x 200mm, for lower basket. Divided into 6 sections, A 13 lid included, mesh size 9 x 9mm
243 mm
1 Each