Learn More
Alfa Aesar™ QuadraSil™ MP, 20-100 micron, metal scavenger
Metal scavenger: Pd (with or without phosphines), Rh, Cu, Ru, Pt, Pb, Ag, Hg
Brand:  Alfa Aesar™ 046259.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15402637
  • 248.57 DKK / 5g
Estimated Shipment date
from Supplier 29-01-2018
Add to basket
Specifications
Chemical Identifiers
QuadraSil™ MP metal scavenger
1225327-73-0
MFCD09753898
QuadraSil™ Mercaptopropyl
Specifications
QuadraSil™ MP metal scavenger
Powder or Beads
QuadraSil™ Mercaptopropyl
White to cream
1225327-73-0
MFCD09753898
20 to 100 Micron
5g