Learn More
Alfa Aesar™ QuadraSil™ AP, 20-100 micron, metal scavenger
Metal scavenger: Co, Cu, Cd, Fe, Ni, Pd, Rh, Ru, Pb, Hg, Zn
Brand:  Alfa Aesar™ 046303.14
Additional Details : Weight : 0.02500kg
Quantity 25g
 View more versions of this product
Product Code. 15402647
  • 796.31 DKK / 25g
Estimated Shipment date
from Supplier 23-01-2018
Add to basket
Specifications
Chemical Identifiers
QuadraSil™ AP metal scavenger
MFCD09753897
QuadraSil™ Aminopropyl
Specifications
QuadraSil™ AP metal scavenger
MFCD09753897
20 to 100 Micron
25g
Powder
QuadraSil™ Aminopropyl