missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QSOX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-14460
Additional Details : Weight : 0.00970kg
Description
QSOX2 Polyclonal antibody specifically detects QSOX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
QSOX2 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.75 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp762A2013, quiescin Q6 sulfhydryl oxidase 2, quiescin Q6-like 1, quiescin Q6-like protein 1 | |
Synthetic 20 amino acid peptide from C-Terminus of human QSOX2. | |
50 μg | |
Cell Biology, Endocrinology, Signal Transduction | |
169714 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |