Learn More
Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes come with either a lid or PTFE cap to limit evaporation.
Brand:  Hellma™ 100-40-40
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10028190
  • 1750.00 DKK / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
  • 80% transmission from 320 to 2500nm
Specifications
Specifications
Macro Absorption Cell
Quartz Suprasil
40 mm
45 mm
14,000 μL
9.5mm
200 to 2500 nm
12.5 mm