missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ qPCR & Go Probe No-ROX Kit
Fast and sensitive qPCR Probe Master Mix. For all other platforms, e.g. Roche, Bio-Rad, Qiagen, etc.
2962.70 DKK - 10213.10 DKK
Specifications
Product Type | qPCR Kit |
---|---|
For Use With (Application) | Real Time PCR (qPCR) |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15848551
|
MP Biomedicals™
11EBI06050 |
500 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15858551
|
MP Biomedicals™
11EBI06200 |
2000 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- The qPCR & Go Probe No-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
- A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR & Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- qPCR & Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Specifications
qPCR Kit | |
Real Time PCR (qPCR) |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.