missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies QNZ
Beskrivelse
- QNZ is a quinazoline derivative that inhibits nuclear factor (NF)-κB activation (IC50 = 11 nM in human Jurkat T lymphocyte cells)
- NF-κB enhances the transcription of pro-inflammatory cytokines, and QNZ inhibits lipopolysaccharide (LPS)-stimulated tumor necrosis factor (TNF)-α production in mouse splenocytes (IC50 = 7 nM;Tobe et al.), as well as CXCL1-mediated pro-inflammatory increase in potassium currents in adult rat neurons (Yang et al)
- It does not inhibit kinases in a standard screen (Wu et al.)
- Neuroprotective in a glutamate toxicity assay using YAC128 medium spiny neuron cultures (Wu et al.)
- Blocks amyloid precursor protein release in human SH-SY5Y neuroblastoma cells caused by muscarinic receptor activation (Choi et al.)
Tekniske data
Tekniske data
| CAS | 545380-34-5 |
| Synonym | CAY10470 |
| Mængde | 1 mg |
| Molekylær formel | C22H20N4O |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?