missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
QKI/Quaking Antibody (PCRP-QKI-2F10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14072H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human samples. It is validated for Western Blot, ELISA
Tekniske data
| QKI/Quaking | |
| Monoclonal | |
| HRP | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| Recombinant full-length human QKI/Quaking protein (Uniprot: Q96PU8) | |
| 0.1 mL | |
| Neuroscience | |
| 9444 | |
| Store at 4°C in the dark. | |
| IgG2c |
| Western Blot, ELISA | |
| PCRP-QKI-2F10 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion