missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 525 Streptavidin Conjugate
Qdot™ 525 Streptavidin Conjugate
Brand: Molecular Probes™ Q10141MP
Additional Details : Weight : 1.05000kg
Description
Qdot™ 525 Streptavidin Conjugate comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Qdot™ nanocrystal). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays, and are also used in purification workflows to achieve targeted fractionation. Qdot™ nanocrystal conjugates are provided as 1μM solutions.Important Features of Qdot™ Streptavidin Conjugates:
Properties of Qdot™ Nanocrystals
The Qdot™ streptavidin conjugate is the size of a large macromolecule or protein (∼15–20 nm) and represents the brightest class of streptavidin detection reagents. Qdot™ streptavidin conjugates are made from a nanometer-scale crystal of a semiconductor material (CdSe), which is coated with an additional semiconductor shell (ZnS) to improve the optical properties of the material. The Qdot™ 705 and Qdot™ 800 streptavidin conjugates, which include CdSeTe, are made in a similar fashion. This core-shell material is further coated with a polymer shell that allows the materials to be conjugated to biological molecules and to retain their optical properties.
Other Fluorescent Conjugates of Streptavidin are Available
We offer several other Qdot™ colors or try the Qdot™ Streptavidin Sampler Kit, containing Qdot™ streptavidin conjugates in six colors. In addition to nanocrystal conjugates, we offer a wide range of streptavidins conjugated to Alexa Fluor™ dyes, Oregon Green™ dye, enzyme conjugates, and traditional fluorophores like Texas Red™ dye, fluorescein (FITC) and more.
Find Biotinylated Conjugates
We offer an extensive line of biotinylated conjugates for use in biotin-streptavidin detection strategies.
Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications
1 μM | |
200 μL | |
Room Temperature | |
Qdot™ |
Qdot 525 | |
Store in refrigerator (2–8°C). | |
Liquid | |
Streptavidin Conjugate (fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.