missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Pyruvic Acid-13C Sodium Salt, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Sodium Pyruvate-3-13C |
| CAS | 124052-04-6 |
| Molekylær formel | 13C C2 H3 O3 . Na |
| Mængde | 500 mg |
| Synonym | Propanoic-3-13C acid','2-oxo-','sodium salt (9CI), Sodium pyruvate-3-13C, Sodium [3-13C]pyruvate, Sodium pyruvate-13C, Sodium 2-oxopropanoate-3-13C, Sodium α-ketopropionate-γ-13C, Pyruvic acid sodium salt-13C, 2-Oxopropanoic-3-13C acid monosodium salt |
| InChI formel | InChI=1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1+1; |
| SMIL | [Na+].[13CH3]C(=O)C(=O)[O-] |
| IUPAC navn | sodium;2-oxo(3^{13}C)propanoate |
| Molekylvægt (g/mol) | 111.037 |
| Formel vægt | 111.001 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?