missing translation for 'onlineSavingsMsg'
Få mere at vide

Pyruvaldehyde-13C3 (2% aqueous solution), TRC

Artikelnummer. 17992365
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17992365 1 mg -
17900925 10 mg -
17980345 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17992365 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9988721MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1353867-77-2
Molekylær formel 13C3H4O2
Molekylvægt (g/mol) 75.04
Synonym Methylglyoxal-13C3
SMIL [13CH3][13C]([13CH]=O)=O

Specifications

Kemisk navn eller materiale Pyruvaldehyde-13C3
Opløsningsmiddel eller Matrix Water
Mængde 1 mg
InChI formel InChI=1S/C3H4O2/c1-3(5)2-4/h2H,1H3/i1+1,2+1,3+1
Formel vægt 75.0312
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.