Få mere at vide
Pyrrolidine-d8, TRC
Artikelnummer. 30423801
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30423801 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423801 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99795225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrrolidine-d8
CAS 212625-79-1
Molekylær formel C4 D8 H N
Mængde 25 mg
Synonym Azacyclopentane-d8, Azolidine-d8, Butylenimine-d8, NSC 62781-d8, Perhydropyrrole-d8, Prolamine-d8, Tetrahydropyrrole-d8, Tetramethylenimine-d8, Pyrrolidine-2,2,3,3,4,4,5,5-d8 (9CI','ACI), Pyrrolidine-D8, Octadeuteriopyrrolidine, Pyrrolidine-2,2,3,3,4,4,5,5-d8
InChI formel InChI=1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2/i1D2,2D2,3D2,4D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 2,2,3,3,4,4,5,5-octadeuteriopyrrolidine
Molekylvægt (g/mol) 79.17
Formel vægt 79.1237