Få mere at vide
Pyrrolidine-2,2,5,5-d4, TRC
Artikelnummer. 30409418
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30409418 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409418 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99795110MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrrolidine-2,2,5,5-D4
CAS 42403-25-8
Molekylær formel C4H5D4N
Mængde 10 mg
Synonym 2,2,5,5-Tetradeuteropyrrolidine
InChI formel InChI=1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2/i3D2,4D2
SMIL [2H]C1([2H])CCC([2H])([2H])N1
IUPAC navn pyrrolidine-2,2,5,5-d4
Molekylvægt (g/mol) 75.15
Formel vægt 75.1