Få mere at vide
Pyriproxyfen-D4, TRC
Artikelnummer. 30420073
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30420073 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420073 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99885210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyriproxyfen-D4
CAS 2446366-95-4
Molekylær formel C20H15D4NO3
Mængde 10 mg
Synonym 2-[1-Methyl-2-(4-phenoxyphenoxy)ethoxy]pyridine-D4
InChI formel InChI=1S/C20H19NO3/c1-16(23-20-9-5-6-14-21-20)15-22-17-10-12-19(13-11-17)24-18-7-3-2-4-8-18/h2-14,16H,15H2,1H3/i5D,6D,9D,14D
SMIL [2H]C1=NC(OC(C)COC(C=C2)=CC=C2OC3=CC=CC=C3)=C([2H])C([2H])=C1[2H]
IUPAC navn 2-((1-(4-phenoxyphenoxy)propan-2-yl)oxy)pyridine-3,4,5,6-d4
Molekylvægt (g/mol) 325.39
Formel vægt 325.16