missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pyrimidine-5-carboxylic acid, 95%

Brand:  Alfa Aesar™ H61406.MD

Additional Details : CAS Number : 4595-61-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15487938

  • 425.85 DKK / 250mg


Estimated Shipment
21-10-2020
Add to basket

Specifications

Chemical Identifiers

4595-61-3
124.099
IIVUJUOJERNGQX-UHFFFAOYSA-N
78346
C1=C(C=NC=N1)C(=O)O
C5H4N2O2
MFCD00856162
5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
pyrimidine-5-carboxylic acid

Specifications

Pyrimidine-5-carboxylic acid
250mg
C5H4N2O2
5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
C1=C(C=NC=N1)C(=O)O
124.099
124.1
95%
259°C to 261°C
4595-61-3
MFCD00856162
IIVUJUOJERNGQX-UHFFFAOYSA-N
pyrimidine-5-carboxylic acid
78346
Solid
Videos
SDS Documents