missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrimidine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-carboxy-Pyrimidine is a synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-carboxy-Pyrimidine is a synthetic intermediate.
Solubility
Soluble in ethanol, Dimethyl sulfoxide (DMSO), Dimethylformamide (DMF), and solution of Phosphate Buffered Saline (PBS).
Notes
Store at -20°C.
2-carboxy-Pyrimidine is a synthetic intermediate.
Solubility
Soluble in ethanol, Dimethyl sulfoxide (DMSO), Dimethylformamide (DMF), and solution of Phosphate Buffered Saline (PBS).
Notes
Store at -20°C.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in ethanol,Dimethyl sulfoxide (DMSO),Dimethylformamide (DMF),and solution of Phosphate Buffered Saline (PBS). |
Formula Weight | 124.1 |
Percent Purity | 97% |
Chemical Name or Material | Pyrimidine-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'