Få mere at vide
Pyrimethanil-d5, TRC
Artikelnummer. 30411490
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30411490 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411490 Leverandør Toronto Research Chemicals Leverandørnr. TRCP997252100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrimethanil phenyl-d5
CAS 2118244-83-8
Renhedsgrad noter HPLC
Molekylær formel C12 2H5 H8 N3
Mængde 100 mg
Synonym Pyrimethanil D5 (phenyl D5)
InChI formel InChI=1S/C12H13N3/c1-9-8-10(2)14-12(13-9)15-11-6-4-3-5-7-11/h3-8H,1-2H3,(H,13,14,15)/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(Nc2nc(C)cc(C)n2)c([2H])c1[2H]
IUPAC navn 4,6-dimethyl-N-(2,3,4,5,6-pentadeuteriophenyl)pyrimidin-2-amine
Molekylvægt (g/mol) 204.28
Formel vægt 204.1423
Procent renhed >95