missing translation for 'onlineSavingsMsg'
Få mere at vide

Pyrimethanil-d5, TRC

Artikelnummer. 30424094
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424094 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424094 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99725210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Pyrimethanil phenyl-d5
CAS 2118244-83-8
Renhedsgrad noter HPLC
Molekylær formel C12 2H5 H8 N3
Mængde 10 mg
Synonym Pyrimethanil D5 (phenyl D5)
InChI formel InChI=1S/C12H13N3/c1-9-8-10(2)14-12(13-9)15-11-6-4-3-5-7-11/h3-8H,1-2H3,(H,13,14,15)/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(Nc2nc(C)cc(C)n2)c([2H])c1[2H]
IUPAC navn 4,6-dimethyl-N-(2,3,4,5,6-pentadeuteriophenyl)pyrimidin-2-amine
Molekylvægt (g/mol) 204.28
Formel vægt 204.1423
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.