missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxine Hydrochloride, Fisher BioReagents
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass
Description
Pyridoxine HCl is used as a vitamin (enzyme cofactor).
Specifications
Melting Point | 214°C |
Color | White |
pH | 2.4 to 3.0 |
Ignition Residue | 0.1% max. |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Conforms to reference |
Assay Percent Range | ≥98 % |
Packaging | Amber Glass |
Quantity | 10 g |
Merck Index | 15, 8095 |
Show More |
Safety and Handling
Product Identifier
- Pyridoxine Hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'