Få mere at vide
Pyridoxine EP Impurity A, Mikromol™
Artikelnummer. 17962056
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17962056 25 mg
17992496 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17962056 Leverandør Mikromol Leverandørnr. MM0021.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pyridoxine EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
CAS 5196-20-3
Analyt- eller komponentnavne 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
Forsendelsestilstand Room Temperature
Molekylær formel C8 H9 N O2
Certificeringer/compliance ISO 17025
Synonym 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol, 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol, Pyridoxine Hydrochloride Imp. A (EP), 1,3-Dihydro-6-methylfuro[3,4-c]pyridin-7-ol, Pyridoxine Imp. A (EP), Pyridoxine Hydrochloride Impurity A, Pyridoxine Impurity A
InChI formel InChI=1S/C8H9NO2/c1-5-8(10)7-4-11-3-6(7)2-9-5/h2,10H,3-4H2,1H3
SMIL Cc1ncc2COCc2c1O
IUPAC navn 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
Molekylvægt (g/mol) 151.16
Mængde 25 mg
Formel vægt 151.0633 g/mol