Få mere at vide
Pyridoxine-d5 (Major), TRC
Artikelnummer. 30428728
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30428728 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428728 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99173710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridoxine-d5
CAS 688302-31-0
Renhedsgrad noter HPLC
Molekylær formel C8H6D5NO3
Mængde 10 mg
Synonym 3,4-Pyridinedimethan-α3,α3-d2-ol','5-hydroxy-6-(methyl-d3)- (9CI), Pyridoxol-d5, 2-(Methyl-d3)-3-hydroxy-4-hydroxymethyl-5-(hydroxymethyl-d2)pyridine, 2-(Methyl-d3)-4-hydroxymethyl-5-(hydroxymethyl-d2)-3-hydroxypyridine, Pyridoxine-d5, Pyridoxine-D5 (2-methyl-D3,5-(hydroxymethyl-D2))
InChI formel InChI=1S/C8H11NO3/c1-5-8(12)7(4-11)6(3-10)2-9-5/h2,10-12H,3-4H2,1H3/i1D3,3D2
SMIL [2H]C([2H])([2H])c1ncc(c(CO)c1O)C([2H])([2H])O
Molekylvægt (g/mol) 174.21
Formel vægt 174.1053
Procent renhed >95