Få mere at vide
Pyridoxine-d2 Hydrochloride, TRC
Artikelnummer. 30427985
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30427985 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427985 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9917341MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridoxine hydrochloride 5-hydroxymethyl-d2
CAS 5027-82-7
Renhedsgrad noter HPLC
Molekylær formel C8 2H2 H9 N O3 . Cl H
Mængde 1 mg
Synonym 3,4-Pyridinedimethan-α3,α3-d2-ol','5-hydroxy-6-methyl-','hydrochloride (9CI), Pyridoxol-α3,α3-d2','hydrochloride (7CI,8CI), Pyridoxol-d2 hydrochloride, 2-Methyl-3-hydroxy-4-hydroxymethyl-5-(hydroxymethyl-d2)pyridine hydrochloride, 2-Methyl-4-hydroxymethyl-5-(hydroxymethyl-d2)-3-hydroxypyridine hydrochloride, Pyridoxine-d2 hydrochloride, Pyridoxine-D2 hydrochloride (5-(hydroxymethyl-D2))
InChI formel InChI=1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H/i3D2;
SMIL Cl.[2H]C([2H])(O)c1cnc(C)c(O)c1CO
IUPAC navn 5-[dideuterio(hydroxy)methyl]-4-(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
Molekylvægt (g/mol) 207.65
Formel vægt 207.0631
Procent renhed >95