Få mere at vide
Pyridoxine-d2 HCl (5-hydroxymethyl-d2), CDN
Artikelnummer. 30315717
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315717 0.01 g
30312508 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315717 Leverandør CDN Leverandørnr. CDND68190.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridoxine hydrochloride 5-hydroxymethyl-d2
CAS 5027-82-7
Molekylær formel C8 2H2 H9 N O3 . Cl H
Mængde 0.01 g
Synonym 3,4-Pyridinedimethan-α3,α3-d2-ol, 5-hydroxy-6-methyl-, hydrochloride (9 CI), Pyridoxol-α3,α3-d2, hydrochloride (7 CI,8 CI), Pyridoxol-d2 hydrochloride, 2-Methyl-3-hydroxy-4-hydroxymethyl-5-(hydroxymethyl-d2)pyridine hydrochloride, 2-Methyl-4-hydroxymethyl-5-(hydroxymethyl-d2)-3-hydroxypyridine hydrochloride, Pyridoxine-d2 hydrochloride, Pyridoxine-D2 hydrochloride (5-(hydroxymethyl-D2))
InChI formel InChI=1 S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12 H,3-4H2,1H3;1 H/i3D2;
SMIL Cl.[2 H]C([2 H])(O)c1cnc(C)c(O)c1CO
IUPAC navn 5-[dideuterio(hydroxy)methyl]-4-(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
Molekylvægt (g/mol) 207.65
Formel vægt 207.0631 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity