Få mere at vide
Pyridoxal-d5 5'-Phosphate, TRC
Artikelnummer. 30415702
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30415702 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415702 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9917170.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridoxal-d5 5'-Phosphate
CAS 1246818-16-5
Renhedsgrad noter HPLC
Molekylær formel C8H5D5NO6P
Mængde 0.5 mg
Synonym 3-Hydroxy-2-methyl-5-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde-d5, Pyridoxal-d5 Phosphate, Pyridoxal-d5 5-(Dihydrogen Phosphate), 2-Methyl-3-hydroxy-4-formyl-5-pyridyl-methylphosphoric Acid-d5, 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde-d5 5-Phosphate, Apolon B6-d5, Biosechs-d5, Codecarboxylase-d5, Coenzyme B6-d5, Hairoxal-d5, Hexermin P-d5, Hi-Pyridoxin-d5, Hiadelon-d5, NSC 82388-d5, PLP-d5, Phosphopyridoxal-d5, Piodel-d5, Pydoxal-d5, Pyridoxal-d5 Monophosphate, Vitazechs-d5
InChI formel InChI=1S/C8H10NO6P/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14/h2-3,11H,4H2,1H3,(H2,12,13,14)/i1D3,4D2
SMIL OC1=C(C([2H])([2H])[2H])N=CC(C([2H])([2H])OP(O)(O)=O)=C1C=O
IUPAC navn [dideuterio-[4-formyl-5-hydroxy-6-(trideuteriomethyl)pyridin-3-yl]methyl] dihydrogen phosphate
Molekylvægt (g/mol) 252.17
Formel vægt 252.056
Procent renhed >95