Få mere at vide
Pyridoxal-d3 Hydrochloride, TRC
Artikelnummer. 30412540
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30412540 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412540 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99172210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridoxal-D3 (methyl-D3) Hydrochloride
CAS 1173023-49-8
Renhedsgrad noter HPLC
Molekylær formel C8 D3 H6 N O3 . HCl
Mængde 10 mg
Synonym 4-Pyridinecarboxaldehyde','3-hydroxy-5-(hydroxymethyl)-2-(methyl-d3)-','hydrochloride (1:1), Pyridoxal-d3 hydrochloride, Pyridoxal-D3 (methyl-D3) hydrochloride, 3-Hydroxy-5-(hydroxymethyl)-2-(methyl-d3)-4-pyridinecarboxaldehyde monohydrochloride, 3-Hydroxy-5-(hydroxymethyl)-2-(trideuteriomethyl)pyridine-4-carboxaldehyde hydrochloride, Pyridoxaldehyde-d3 hydrochloride
InChI formel InChI=1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])c1ncc(CO)c(C=O)c1O
IUPAC navn 3-hydroxy-5-(hydroxymethyl)-2-(trideuteriomethyl)pyridine-4-carbaldehyde;hydrochloride
Molekylvægt (g/mol) 206.641
Formel vægt 206.0538
Procent renhed >95