Få mere at vide
Pyridiniumdichromat, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Pyridinium Dichromate |
| CAS | 20039-37-6 |
| Molekylær formel | C10H12Cr2N2O7 |
| Mængde | 1 g |
| Synonym | Pyridine Compd. with Dichromic Acid, Dichromic Acid Compd. with Pyridine, Pyridine Compd. with Chromic Acid, Bis(pyridinium) Dichromate, Dipyridinium Dichromate, PDC |
| InChI formel | InChI=1S/2C5H5N.2Cr.2H2O.5O/c2*1-2-4-6-5-3-1;;;;;;;;;/h2*1-5H;;;2*1H2;;;;;/q;;2*+1;;;;;;;/p-2 |
| SMIL | O[Cr](=O)(=O)O[Cr](=O)(=O)O.c1ccncc1.c2ccncc2 |
| IUPAC navn | hydroxy-(hydroxy(dioxo)chromio)oxy-dioxochromium; Pyridin |
| Molekylvægt (g/mol) | 376.2 |
| Formel vægt | 375.9455 |