Learn More
Pyridine-3,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-7081529
Click to view available options
Quantity:
10 g
50 g
499-81-0
C7H5NO4
167.12
MFCD00006393
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f
10366
CHEBI:46875
pyridine-3,5-dicarboxylic acid
OC(=O)C1=CC(=CN=C1)C(O)=O
499-81-0
C7H5NO4
167.12
MFCD00006393
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f
10366
CHEBI:46875
pyridine-3,5-dicarboxylic acid
OC(=O)C1=CC(=CN=C1)C(O)=O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point ∼325°C (decomposition)
Quantity 10 g
Beilstein 131640
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material Pyridine-3,5-dicarboxylic acid
compliance-icons
Product Identifier
  • Pyridine-3,5-dicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Pyridine-3,5-dicarboxylic acid