Få mere at vide
Pyridine-2-carboxaldehyde Methiodide, TRC
Artikelnummer. 30419483
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419483 1 g
30410144 100 mg
30414373 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30419483 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9918031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3784-97-2
Molekylær formel C7H8INO
Molekylvægt (g/mol) 249.05
Synonym 2-Formyl-1-methylpyridinium Iodide, PCAM
IUPAC navn 1-methylpyridin-1-ium-2-carbaldehyde;iodide
SMIL O=CC1=[N+](C)C=CC=C1.[I-]
Tekniske data
Kemisk navn eller materiale Pyridine-2-carboxaldehyde Methiodide
Mængde 1 g
InChI formel InChI=1S/C7H8NO.HI/c1-8-5-3-2-4-7(8)6-9;/h2-6H,1H3;1H/q+1;/p-1
Formel vægt 248.9651