Få mere at vide
Pyridine-2,6-d2, TRC
Artikelnummer. 30419240
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30419240 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419240 Leverandør Toronto Research Chemicals Leverandørnr. TRCP998912100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyridine-2,6-D2
CAS 17265-96-2
Molekylær formel C5H3D2N
Mængde 100 mg
Synonym Pyridine-2,6-d2 (6CI','7CI','8CI','9CI','ACI), 2,6-Dideuteropyridine, Pyridine-2,6-D2
InChI formel InChI=1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i4D,5D
SMIL [2H]C1=CC=CC([2H])=N1
IUPAC navn pyridine-2,6-d2
Molekylvægt (g/mol) 81.11
Formel vægt 81.05