Learn More
Pyrex™ Wide Neck Round Bottom Boiling Flask
Made of borosilicate glass
Brand: Pyrex 1120/14D
1707.55 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Complies with ISO 24450 and DIN 12353
- With wide neck to facilitate addition of solid samples etc
- Resistant to chemical corrosion and thermal shock
Specifications
ISO 24450, DIN 12353 | |
Borosilicate Glass | |
Wide |
500 mL | |
168 mm | |
Round |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
Pyrex™ Wide Neck Round Bottom Boiling Flask > Capacity: 500mL
Spot an opportunity for improvement?Share a Content Correction