Learn More
Pyrex™ Borosilicate Glass Watch Glasses
Product Code. 10681101
Change view
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
80 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Unit Size:
Each
Pack of 10
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10681101 40 mm Pack of 10
11303454 100 mm Each
10468370 100 mm Pack of 10
10651651 125 mm Pack of 10
11313454 125 mm Each
10315100 150 mm Pack of 10
11323454 150 mm Each
11367824 250 mm Each
10101780 250 mm Pack of 10
11373444 50 mm Each
10706071 50 mm Pack of 10
11363444 40 mm Each
10706261 60 mm Pack of 10
11383444 60 mm Each
11926288 80 mm Pack of 10
11393444 80 mm Each
13481829 200 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 10681101 Supplier Pyrex Supplier No. 1060/04D
  • ONLINE EXCLUSIVE PRICE
    1225.00 DKK
    1057.10 DKK / Pakke med 10
    Spare 167.90 DKK
    13% Slukket
Forventet forsendelse: 08-03-2027
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex™ borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Specifications
Bottom Shape Flat
Material Borosilicate Glass
Shape Round
Type Watch Glass
For Use With (Application) Evaporation Work and Covering Heated Beakers
Interior Finish Polished
Diameter (Metric) 40 mm