missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™175mm Diameter Glass Büchner Filter Funnel

Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 175mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.

Brand:  Pyrex 3722/04M

2489.00 DKK valid until 2024-12-31
Use promo code "23259" to get your promotional price.



Additional Details : Weight : 1.83000kg

 View more versions of this product

Product Code. 12858180

  • 2620.00 DKK / Pack of 2
Estimated Shipment: 04-11-2024
to see stock.
Add to basket

Description

Description

  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions
    - No filter papers required
    - Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Choice of disc diameters and porosities
Specifications

Specifications

Buchner filter funnel
30mm
390 mm
175mm
150mm
40 to 100μm
4000 mL
Pyrex Glass, Borosilicate Glass
For easy filtering solutions
Integrated filter plate sintered Glass disc
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers