Få mere at vide
Pyrethrum Extract (Technical Grade), TRC
Artikelnummer. 17971485
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17971485 5 mg
17981185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17971485 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9912485MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 8003-34-7
Molekylær formel C43H56O8
Synonym Pyrethrins, Pyrethrins and Pyrethroids, Biopiren plus, Chrysanthemates pyrethrins and pyrethroids, Diatect, Firmotox, Goldgeist forte, Nettafid, Parexan, Piresan, Pyganic, Pyganic MUP 20, Pyre-sin Plus, Pyrellin, Pyrethrine, Pyrethrines, Pyrethrins and Pyrethroids, chrysanthemates, Pyrethrins and Pyrethroids, esters, Pyrethrins, chrysanthemates, Pyrethro Vioryl 5SC, Pyrethrum, Pyrethrum (insecticide), Pyrethrum FS, Pyrevert, Pyrocide 50, Spruzit, Take Down, Take Down Garden Spray
SMIL C/C(C)=C/C1C(C)(C)[C@@H]1C(O[C@@H]2C(C)=C(C/C=C/C=C)C(C2)=O)=O.C/C(C([CH3]=O)=O)=C\C3C(C)(C)[C@@H]3C(OC4C(C)=C(C/C=C/C=C)C(C4)=O)=O
Tekniske data
Kemisk navn eller materiale Pyrethrins
Mængde 5 mg
InChI formel InChI=1S/C22H28O5.C21H28O3/c1-7-8-9-10-15-14(3)18(12-17(15)23)27-21(25)19-16(22(19,4)5)11-13(2)20(24)26-6;1-7-8-9-10-15-14(4)18(12-17(15)22)24-20(23)19-16(11-13(2)3)21(19,5)6/h7-9,11,16,18-19H,1,10,12H2,2-6H3;7-9,11,16,18-19H,1,10,12H2,2-6H3/b9-8+,13-11+;9-8+/t16?,18?,19-;16?,18-,19-/m00/s1