Få mere at vide
Pyrene-d10, TRC
Artikelnummer. 30423554
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30423554 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423554 Leverandør Toronto Research Chemicals Leverandørnr. TRCP849002100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrene-d10
CAS 1718-52-1
Renhedsgrad noter HPLC
Molekylær formel C16 2H10
Mængde 100 mg
Synonym Pyrene-1,2,3,4,5,6,7,8,9,10-d10, Pyrene-d10 (7CI,8CI,9CI), Perdeuteriopyrene, [10-2H]Pyrene, [2H10]Pyrene, Pyrene D10
InChI formel InChI=1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c2c([2H])c([2H])c3c([2H])c([2H])c([2H])c4c([2H])c([2H])c(c1[2H])c2c34
IUPAC navn 1,2,3,4,5,6,7,8,9,10-decadeuteriopyrene
Molekylvægt (g/mol) 212.31
Formel vægt 212.141
Procent renhed >95