Få mere at vide
Pyrazolo[1,5-a]pyrimidine-3-thiocarboxamide, 97%, Thermo Scientific™
Artikelnummer. 15424568
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
Artikelnummer. 15424568
Brand: Thermo Scientific Alfa Aesar H58241.03
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1421263-27-5
Molekylær formel C7H6N4S
Molekylvægt (g/mol) 178.213
MDL nummer MFCD22682952
InChI nøgle SQCMCBNIRYVBTG-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-3-carbothioamide, pyrazolo 1,5-a pyrimidine-3-thiocarboxamide
PubChem CID 86277501
IUPAC navn pyrazolo[1,5-a]pyrimidine-3-carbothioamide
SMIL C1=CN2C(=C(C=N2)C(=S)N)N=C1
Tekniske data
Kemisk navn eller materiale Pyrazolo[1,5-a]pyrimidine-3-thiocarboxamide
Mængde 1 g
Formel vægt 178.21
Procent renhed 97%