Få mere at vide
Pyrazinecarboxylic Acid-d3, TRC
Artikelnummer. 30427472
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30427472 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427472 Leverandør Toronto Research Chemicals Leverandørnr. TRCP84075210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrazinecarboxylic Acid-d3
CAS 1794791-32-4
Molekylær formel C5 D3 H N2 O2
Mængde 10 mg
Synonym 3,5,6-Trideuteriopyrazine-2-carboxylic acid, Pyrazine-2-carboxylic Acid-3,5,6-D3, 2-Pyrazine-d3-carboxylic Acid, Pyrazinecarboxylic Acid-d3
InChI formel InChI=1S/C5H4N2O2/c8-5(9)4-3-6-1-2-7-4/h1-3H,(H,8,9)/i1D,2D,3D
SMIL [2H]c1nc([2H])c(nc1[2H])C(=O)O
IUPAC navn 3,5,6-trideuteriopyrazine-2-carboxylic acid
Molekylvægt (g/mol) 127.116
Formel vægt 127.0461