missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinecarbonitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-4259062
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19847-12-2
C5H3N3
105.10
MFCD00049361
PMSVVUSIPKHUMT-UHFFFAOYSA-N
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
73172
CHEBI:3982
pyrazine-2-carbonitrile
N#CC1=CN=CC=N1
This item is not returnable. View return policy
19847-12-2
C5H3N3
105.10
MFCD00049361
PMSVVUSIPKHUMT-UHFFFAOYSA-N
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
73172
CHEBI:3982
pyrazine-2-carbonitrile
N#CC1=CN=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 20°C
Color Colorless to Yellow
Density 1.1740g/mL
Boiling Point 90°C to 93°C (15.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5330 to 1.5350 (20°C, 589nm)
Quantity 10 g
Specific Gravity 1.174
Solubility Information Solubility in water: soluble
Formula Weight 105.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Pyrazinecarbonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Pyrazinecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'