Få mere at vide
Pyrazine-d4, TRC
Artikelnummer. 30413543
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30413543 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413543 Leverandør Toronto Research Chemicals Leverandørnr. TRCP840747250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrazine-D4
CAS 1758-62-9
Molekylær formel C4 D4 N2
Mængde 250 mg
Synonym Pyrazine-2,3,5,6-D4, Perdeuteriopyrazine, 1,4-Diazabenzene-D4, 1,4-Diazine-D4, Nsc 400221-D4, Paradiazine-D4, Piazine-D4, P-Diazine-D4
InChI formel InChI=1S/C4H4N2/c1-2-6-4-3-5-1/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c([2H])nc1[2H]
IUPAC navn 2,3,5,6-tetradeuteriopyrazine
Molekylvægt (g/mol) 84.06
Formel vægt 84.1126