Få mere at vide
Pyrazine-d4, CDN
Artikelnummer. 30319551
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319551 0.5 g 0.5g
30316553 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319551 Leverandør CDN Leverandørnr. CDND00840.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrazine-D4
CAS 1758-62-9
Molekylær formel C4 D4 N2
Mængde 0.5 g
Synonym Pyrazine-2,3,5,6-D4, Perdeuteriopyrazine, 1,4-Diazabenzene-D4, 1,4-Diazine-D4, Nsc 400221-D4, Paradiazine-D4, Piazine-D4, P-Diazine-D4
InChI formel InChI=1 S/C4H4N2/c1-2-6-4-3-5-1/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1nc([2 H])c([2 H])nc1[2 H]
IUPAC navn 2,3,5,6-tetradeuteriopyrazine
Molekylvægt (g/mol) 84.06
Formel vægt 84.1126 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity