Få mere at vide
Pyrazinamide-15N,d3, TRC
Artikelnummer. 30419410
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30419410 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419410 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8406021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pyrazinamide-15N,D3
CAS 1246817-81-1
Renhedsgrad noter HPLC
Molekylær formel C5H2D3N215NO
Mængde 1 mg
Synonym Pyrazinamide-(amide-15N,3,5,6-D3), Pyrazinamide-(3,5,6-d3,amide-15N), Pyrazin-3,5,6-D3-amide-15N, Pyrazinamide-15N,d3, 2-Pyrazine-3,5,6-d3-carboxamide-15N, 2-(Amino-15N-carbonyl)pyrazine-3,5,6-d3
InChI formel InChI=1S/C5H5N3O/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)/i1D,2D,3D,6+1
SMIL [2H]c1nc([2H])c(nc1[2H])C(=O)[15NH2]
Molekylvægt (g/mol) 127.12
Formel vægt 127.0591
Procent renhed >95