Få mere at vide
Pyrathiazine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pyrathiazine Hydrochloride |
| CAS | 522-25-8 |
| Analyt- eller komponentnavne | Pyrathiazine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H20 N2 S . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine, 10-[2-(1-pyrrolidinyl)ethyl]-, hydrochloride (1:1), 10H-Phenothiazine, 10-[2-(1-pyrrolidinyl)ethyl]-, monohydrochloride (9CI), Phenothiazine, 10-[2-(1-pyrrolidinyl)ethyl]-, hydrochloride (7CI), Phenothiazine, 10-[2-(1-pyrrolidinyl)ethyl]-, monohydrochloride (8CI), 10-[2-(1-Pyrrolidyl)ethyl]phenothiazine hydrochloride, N-(2-Pyrrolidinoethyl)phenothiazine hydrochloride, Parathiazine hydrochloride, Pyrathiazine hydrochloride, Pyrrolazote, Pyrrolazote abergic, Pyrrolazote hydrochloride, Rolazote |
| InChI formel | InChI=1S/C18H20N2S.ClH/c1-3-9-17-15(7-1)20(14-13-19-11-5-6-12-19)16-8-2-4-10-18(16)21-17;/h1-4,7-10H,5-6,11-14H2;1H |
| Løsningstype | Neat |
| SMIL | Cl.C(CN1c2ccccc2Sc3ccccc13)N4CCCC4 |
| Vis mere |