Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pyraclostrobin |
| CAS | 175013-18-0 |
| CAS Max % | ≤ 100% |
| Smeltepunkt | 64°C |
| Kogepunkt | 200°C (Decomposition) |
| Synonym | Methyl {2-[1-(4-chlorophenyl)-1H-pyrazol-3-yloxymethyl]phenyl}methoxycarbamate |
| Molekylvægt (g/mol) | 387.82 g/mol |
| Mængde | 100 mg |