missing translation for 'onlineSavingsMsg'
Få mere at vide

Pyr-Gly-OH, TRC

Artikelnummer. 30419413
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419413 100 mg
30418612 50 mg
30410015 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30419413 Leverandør Toronto Research Chemicals Leverandørnr. TRCP841188100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29227-88-1
Molekylær formel C7H10N2O4
Molekylvægt (g/mol) 186.17
Synonym Pyroglutamylglycine
IUPAC navn 2-[[(2S)-5-oxopyrrolidine-2-carbonyl]amino]acetic acid
SMIL OC(=O)CNC(=O)[C@@H]1CCC(=O)N1

Specifications

Kemisk navn eller materiale Pyr-Gly-OH
InChI formel InChI=1S/C7H10N2O4/c10-5-2-1-4(9-5)7(13)8-3-6(11)12/h4H,1-3H2,(H,8,13)(H,9,10)(H,11,12)/t4-/m0/s1
Mængde 100 mg
Formel vægt 186.0641
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.