Få mere at vide
Pyr-Gly-OH, TRC
Artikelnummer. 30418612
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418612 50 mg
30410015 10 mg
30419413 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30418612 Leverandør Toronto Research Chemicals Leverandørnr. TRCP84118850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 29227-88-1
Molekylær formel C7H10N2O4
Molekylvægt (g/mol) 186.17
Synonym Pyroglutamylglycine
IUPAC navn 2-[[(2S)-5-oxopyrrolidine-2-carbonyl]amino]acetic acid
SMIL OC(=O)CNC(=O)[C@@H]1CCC(=O)N1
Tekniske data
Kemisk navn eller materiale Pyr-Gly-OH
InChI formel InChI=1S/C7H10N2O4/c10-5-2-1-4(9-5)7(13)8-3-6(11)12/h4H,1-3H2,(H,8,13)(H,9,10)(H,11,12)/t4-/m0/s1
Mængde 50 mg
Formel vægt 186.0641