Learn More
Lenz Laborglasintrumente Pycnometers acc. To Gay Lussac Unadjusted
Product Code. 10797701
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10797701 5 mL Each
10315691 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10797701 Supplier Lenz Laborglasintrumente Supplier No. 05630010
  • ONLINE EXCLUSIVE PRICE
    255.00 DKK
    205.23 DKK / Pakke med 1
    Spare 49.77 DKK
    19% Slukket
Forventet forsendelse: 24-02-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Gay Lussac pycnometers acc. to DIN ISO 3507
  • Unadjusted, stopper NS 10/19
  • With capillary, stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications
Capacity (Metric) 5 mL
Material Borosilicate Glass (3.3)