Learn More
Lenz Laborglasintrumente Pycnometers Acc. to Gay Lussac Adjusted
Product Code. 10385781
Change view
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
100 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10385781 25 mL Each
10400903 5 mL Each
10778271 50 mL Each
10335881 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10385781 Supplier Lenz Laborglasintrumente Supplier No. 05630222
  • ONLINE EXCLUSIVE PRICE
    637.00 DKK
    556.95 DKK / Pakke med 1
    Spare 80.05 DKK
    12% Slukket
View all promotions
Forventet forsendelse: 28-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Gay Lussac pycnometers acc. to DIN ISO 3507
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Adjusted to In (with individual certificate)
  • Accuracy: 0.001 cm³
  • Material: borosilicate glass 3.3
  • Capcity shown is nominal capacity
Specifications
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)