missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PVRL1 Monoclonal Antibody (OTI6F8), TrueMAB™, OriGene
Beskrivelse
This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
Tekniske data
Tekniske data
| Antigen | PVRL1 |
| Applikationer | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI6F8 |
| Koncentration | 0.54 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | NECTIN1 |
| Genadgangsnr. | Q15223 |
| Gene Alias | CD111, CLPED1, ED4, HIgR, HV1S, HVEC, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK-12, nectin-1 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?